9-Trifluoroacetylanthracene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
9-Anthryl Trifluoromethyl Ketone
PC4531-1G
PC4531-5G
APOSPC4531-1GEA
87.3
EUR
APOSPC4531-1G
APOSPC4531-5G
9-Trifluoroacetylanthracene
9-Trifluoroacetylanthracene
Formel:
C₁₆H₉F₃O Molecular Weight: 274.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001258 CAS-Nummer: 53531-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...