1H,1H,9H-Hexadecafluorononyl methacrylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H,1H,9H-Perfluorononyl methacrylate
PC4535-25G
PC4535-5G
APOSPC4535-25GEA
140
EUR
APOSPC4535-25G
APOSPC4535-5G
1H,1H,9H-Hexadecafluorononyl methacrylate 97%
1H,1H,9H-Hexadecafluorononyl methacrylate
Formel:
C₁₃H₈F₁₆O₂ Molecular Weight: 500,18 g/mol Siedepunkt: 234 °C Dichte: 1,618 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078427 CAS-Nummer: 1841-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...