5-Fluor-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC4537-10G PC4537-5G PC4537-1G
APOSPC4537-10GEA 52.2 EUR
APOSPC4537-10G APOSPC4537-5G APOSPC4537-1G
5-Fluor-2-(trifluormethyl)benzylbromid 98%
5-Fluor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00236290
CAS-Nummer: 239135-48-9
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR