2-Fluor-4-iodbenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4561-1G
PC4561-5G
APOSPC4561-1GEA
53.8
EUR
APOSPC4561-1G
APOSPC4561-5G
2-Fluor-4-iodbenzotrifluorid 98%
2-Fluor-4-iodbenzotrifluorid
Formel:
C₇H₃F₄I Lagertemperatur: Kühlschrank |
MDL:
MFCD01320764 CAS-Nummer: 239135-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...