4-(Trifluoromethyl)-DL-phenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-Phe(4-CF₃)-OH

PC4562-100MG PC4562-1G PC4562-5G
APOSPC4562-100MGEA 29 EUR
APOSPC4562-100MG APOSPC4562-1G APOSPC4562-5G
4-(Trifluoromethyl)-DL-phenylalanine 97%
4-(Trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233.19 g/mol
Schmelzpunkt: 200…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070813
CAS-Nummer: 14091-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR