2,3,4,5-Tetrafluor-6-nitrobenzoesäure

Lieferant: Apollo Scientific
Achtung

PC4583-1G PC4583-500MG PC4583-5G
APOSPC4583-1GEA 36.4 EUR
APOSPC4583-1G APOSPC4583-500MG APOSPC4583-5G
2,3,4,5-Tetrafluor-6-nitrobenzoesäure
2,3,4,5-Tetrafluor-6-nitrobenzoesäure
Formel: C₇HF₄NO₄
Molecular Weight: 239,08 g/mol
Schmelzpunkt: 137…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093916
CAS-Nummer: 16583-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR