α,α,α,5-Tetrafluor-o-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4609-100G
PC4609-25G
PC4609-500G
APOSPC4609-100GEA
319
EUR
APOSPC4609-100G
APOSPC4609-25G
APOSPC4609-500G
α,α,α,5-Tetrafluor-o-toluylsäure 98%
α,α,α,5-Tetrafluor-o-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208,11 g/mol Schmelzpunkt: 78…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083521 CAS-Nummer: 654-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...