1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
O-[3-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride
PC4629-1G
PC4629-500MG
APOSPC4629-1GEA
132
EUR
APOSPC4629-1G
APOSPC4629-500MG
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
Formel:
C₈H₉ClF₃NO Schmelzpunkt: 160…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01114571 CAS-Nummer: 15256-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...