2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile 97%

Lieferant: Apollo Scientific

PC4633-1G PC4633-500MG PC4633-5G
APOSPC4633-1GEA 172 EUR
APOSPC4633-1G APOSPC4633-500MG APOSPC4633-5G
2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile 97%
2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile
Formel: C₂₁H₁₄FNO
Molecular Weight: 315.35 g/mol
Schmelzpunkt: 89…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01316984
CAS-Nummer: 446275-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR