4-Chlor-3-(trifluormethyl)benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlor-α,α,α-trifluor-m-toluylsäure
PC4634-1G
PC4634-25G
PC4634-5G
APOSPC4634-1GEA
21.9
EUR
APOSPC4634-1G
APOSPC4634-25G
APOSPC4634-5G
4-Chlor-3-(trifluormethyl)benzoesäure 97%
4-Chloro-3-(trifluoromethyl)benzoic acid
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224,57 g/mol Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631468 CAS-Nummer: 1737-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...