1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
1-[3,5-Bis(trifluoromethyl)benzyl]pyrid-2-one-3-carboxylic acid
PC4635-1G
PC4635-500MG
APOSPC4635-1GEA
799
EUR
APOSPC4635-1G
APOSPC4635-500MG
1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 97%
1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formel:
C₁₅H₉F₆NO₃ Molecular Weight: 365.23 g/mol Schmelzpunkt: 168…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243697 CAS-Nummer: 338781-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...