N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide 97%

Lieferant: Apollo Scientific

PC4636-1G PC4636-500MG PC4636-5G
APOSPC4636-1GEA 172 EUR
APOSPC4636-1G APOSPC4636-500MG APOSPC4636-5G
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide 97%
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide
Formel: C₁₃H₁₂ClF₆NO
Molecular Weight: 347,69 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793437
CAS-Nummer: 478261-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR