2,2,3,4,4,4-Hexafluorbutylmethacrylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,3,4,4,4-Hexafluorobutyl Methacrylate
PC4657-100G
PC4657-25G
PC4657-5G
APOSPC4657-100GEA
241
EUR
APOSPC4657-100G
APOSPC4657-25G
APOSPC4657-5G
2,2,3,4,4,4-Hexafluorbutylmethacrylat 98%
2,2,3,4,4,4-Hexafluorbutylmethacrylat
Stabilised; Refrigeration will extend shelf-life.
Formel:
C₈H₈F₆O₂ Molecular Weight: 250,14 g/mol Siedepunkt: 158 °C Dichte: 1,348 g/cm³ (25 °C) Flammpunkt: 57 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042311 CAS-Nummer: 36405-47-7 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...