4-Chloro-6,8-difluoroquinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC4675-1G
PC4675-250MG
APOSPC4675-1GEA
130
EUR
APOSPC4675-1G
APOSPC4675-250MG
4-Chloro-6,8-difluoroquinoline
4-Chloro-6,8-difluoroquinoline
Formel:
C₉H₄ClF₂N Molecular Weight: 199,585 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00278782 CAS-Nummer: 239463-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...