4-Chloro-1,1,2-trifluoro-1-butene 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4727-1G
PC4727-25G
PC4727-5G
APOSPC4727-1GEA
87
EUR
APOSPC4727-1G
APOSPC4727-25G
APOSPC4727-5G
4-Chloro-1,1,2-trifluoro-1-butene 97%
4-Chloro-1,1,2-trifluoro-1-butene
Formel:
C₄H₄ClF₃ Molecular Weight: 144,52 g/mol Dichte: 1,25 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077488 CAS-Nummer: 235106-11-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...