Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester
PC4732-100G
PC4732-25G
APOSPC4732-100GEA
292
EUR
APOSPC4732-100G
APOSPC4732-25G
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Formel:
C₉H₁₁F₃O₄ Molecular Weight: 240,18 g/mol Siedepunkt: 80…82 °C/1mmHg Dichte: 1,235 g/cm³ (25 °C) Flammpunkt: 104 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02677683 CAS-Nummer: 571-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...