2-Fluorphenylacetat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Acetoxy-2-fluorbenzol, Essigsäure(2-fluorphenyl)ester
PC4739-1G
PC4739-25G
PC4739-5G
APOSPC4739-1GEA
169
EUR
APOSPC4739-1G
APOSPC4739-25G
APOSPC4739-5G
2-Fluorphenylacetat 98%
2-Fluorphenylacetat
Formel:
C₈H₇FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672912 CAS-Nummer: 29650-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...