4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane

PC4755-5G PC4755-100G PC4755-25G
APOSPC4755-5GEA 21.9 EUR
APOSPC4755-5G APOSPC4755-100G APOSPC4755-25G
4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid 99%
4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid

Intermediate for high performance polyimides.

Formel: C₈H₄O₃
Molecular Weight: 444.25 g/mol
Schmelzpunkt: 244…247 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039143
CAS-Nummer: 1107-00-2
EINECS: 214-170-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR