5-Brom-2,3-difluoranilin

Lieferant: Apollo Scientific
Achtung

PC47692-1G PC47692-5G
APOSPC47692-1GEA 348 EUR
APOSPC47692-1G APOSPC47692-5G
5-Brom-2,3-difluoranilin
5-Brom-2,3-difluoranilin
Formel: C₆H₄BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD22205725
CAS-Nummer: 1375068-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR