2-Brom-5-fluorisonicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-fluoro-4-pyridinecarboxylic acid, 2-Bromo-5-fluoropyridine-4-carboxylic acid
PC48293-1G
PC48293-5G
PC48293-25G
APOSPC48293-1GEA
21.9
EUR
APOSPC48293-1G
APOSPC48293-5G
APOSPC48293-25G
2-Brom-5-fluorisonicotinsäure 97%
2-Brom-5-fluorisonicotinsäure
Formel:
C₆H₃BrFNO₂ Molecular Weight: 219.9959 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261246 CAS-Nummer: 885588-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...