2-Chlor-6-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Brommethyl)-2-chlor-6-(trifluormethyl)benzol

PC48302-25G PC48302-5G PC48302-100G
APOSPC48302-25GEA 279 EUR
APOSPC48302-25G APOSPC48302-5G APOSPC48302-100G
2-Chlor-6-(trifluormethyl)benzylbromid 98%
2-Chlor-6-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Molecular Weight: 273.48 g/mol
Dichte: 1.6951 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD06660232
CAS-Nummer: 886500-26-1
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR