4-(2-Fluoro-1,1'-biphenyl-4-yl)pyrimidin-2-amine

Lieferant: Apollo Scientific

PC48315-1G PC48315-250MG
APOSPC48315-1GEA 401 EUR
APOSPC48315-1G APOSPC48315-250MG
4-(2-Fluoro-1,1'-biphenyl-4-yl)pyrimidin-2-amine
4-(2-Fluoro-1,1'-biphenyl-4-yl)pyrimidin-2-amine
Formel: C₁₆H₁₂FN₃
Molecular Weight: 265.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089178
CAS-Nummer: 1287217-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR