5-Fluor-2-methoxy-4-pyrimidinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-methoxypyrimidin-4(3H)-one
PC48339-100G
PC48339-25G
PC48339-5G
APOSPC48339-100GEA
58
EUR
APOSPC48339-100G
APOSPC48339-25G
APOSPC48339-5G
5-Fluor-2-methoxy-4-pyrimidinol
5-Fluor-2-methoxy-4-pyrimidinol
Formel:
C₅H₅FN₂O₂ Molecular Weight: 144.11 g/mol Schmelzpunkt: 200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03265437 CAS-Nummer: 1480-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...