5-Methoxy-2-fluorobenzylcyanide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-methoxyphenylacetonitrile

PC48340-1G PC48340-5G PC48340-250MG
APOSPC48340-1GEA 94.5 EUR
APOSPC48340-1G APOSPC48340-5G APOSPC48340-250MG
5-Methoxy-2-fluorobenzylcyanide 97%
5-Methoxy-2-fluorobenzylcyanide
Formel: C9H8FNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 672931-28-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR