2-(Difluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC48345-1G PC48345-250MG PC48345-5G
APOSPC48345-1GEA 43.5 EUR
APOSPC48345-1G APOSPC48345-250MG APOSPC48345-5G
2-(Difluoromethyl)benzoic acid 97%
2-(Difluoromethyl)benzoic acid
Formel: C₈H₆F₂O₂
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140191
CAS-Nummer: 799814-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR