2-Methoxy-3-(trifluormethyl)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC48351-1G PC48351-25G PC48351-5G
APOSPC48351-1GEA 48 EUR
APOSPC48351-1G APOSPC48351-25G APOSPC48351-5G
2-Methoxy-3-(trifluormethyl)benzoesäure 98%
2-Methoxy-3-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832311
CAS-Nummer: 937068-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR