2-Chloro-6-(trifluoromethyl)pyrazine

Lieferant: Apollo Scientific
Gefahr

PC48361-10G PC48361-1G PC48361-5G
APOSPC48361-10GEA 1070 EUR
APOSPC48361-10G APOSPC48361-1G APOSPC48361-5G
2-Chloro-6-(trifluoromethyl)pyrazine
2-Chloro-6-(trifluoromethyl)pyrazine
Formel: C₅H₂ClF₃N₂
Molecular Weight: 182.53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189477
CAS-Nummer: 61655-69-4
UN: 3267
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR