2-(Difluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC48438-1G PC48438-250MG PC48438-5G
APOSPC48438-1GEA 0 EUR
APOSPC48438-1G APOSPC48438-250MG APOSPC48438-5G
2-(Difluoromethyl)benzaldehyde 95%
2-(Difluoromethyl)benzaldehyde
Formel: C₈H₆F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16875639
CAS-Nummer: 1018678-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR