6-Methyl-2-(trifluoromethyl)quinolin-4-ol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline

PC4858E-1G PC4858E-5G
APOSPC4858E-1GEA 29.2 EUR
APOSPC4858E-1G APOSPC4858E-5G
6-Methyl-2-(trifluoromethyl)quinolin-4-ol 97%
6-Methyl-2-(trifluoromethyl)quinolin-4-ol
Formel: C₁₁H₈F₃NO
Molecular Weight: 227,19 g/mol
Schmelzpunkt: 252…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153192
CAS-Nummer: 1701-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR