2,5-Difluorobenzeneboronic acid pinacol ester

Lieferant: Apollo Scientific

Synonyme: 2,5-Difluorophenylboronic acid pinacol ester, 2-(2,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC48624-1G
APOSPC48624-1GEA 119 EUR
APOSPC48624-1G
2,5-Difluorobenzeneboronic acid pinacol ester
2,5-Difluorobenzeneboronic acid pinacol ester
Formel: C₁₂H₁₅BF₂O₂
Molecular Weight: 240,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405346
CAS-Nummer: 408492-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR