Indium(III)fluorid, wasserfrei 99%

Lieferant: Apollo Scientific
Achtung

PC4870-100G PC4870-25G
APOSPC4870-100GEA 865 EUR
APOSPC4870-100G APOSPC4870-25G
Indium(III)fluorid, wasserfrei 99%
Indium(III)fluorid
Formel: F₃In
Molecular Weight: 171,82 g/mol
Schmelzpunkt: 1170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016149
CAS-Nummer: 7783-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR