Indium(III) trifluoroacetate 98%

Lieferant: Apollo Scientific
Achtung

PC4872-1G PC4872-5G
APOSPC4872-1GEA 109 EUR
APOSPC4872-1G APOSPC4872-5G
Indium(III) trifluoroacetate 98%
Indium(III) trifluoroacetate
Lagertemperatur: Raumtemperatur
MDL: MFCD00050940
CAS-Nummer: 36554-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR