3-Fluor-5-nitrobenzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC48725-5G
APOSPC48725-5GEA
95.7
EUR
APOSPC48725-5G
3-Fluor-5-nitrobenzoesäure 97%
3-Fluor-5-nitrobenzoesäure
Formel:
C₇H₄FNO₄ Molecular Weight: 185,11 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD05864360 CAS-Nummer: 14027-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...