4-(4-(Trifluormethyl)phenoxy)anilin

Lieferant: Apollo Scientific
Achtung

PC48729-1G PC48729-5G
APOSPC48729-1GEA 190 EUR
APOSPC48729-1G APOSPC48729-5G
4-(4-(Trifluormethyl)phenoxy)anilin
4-(4-(Trifluormethyl)phenoxy)anilin
Formel: C₁₃H₁₀F₃NO
Molecular Weight: 253.220 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00170930
CAS-Nummer: 57478-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR