2'-Desoxy-2'-fluoruridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
PC48732-10G
PC48732-25G
PC48732-5G
APOSPC48732-10GEA
132
EUR
APOSPC48732-10G
APOSPC48732-25G
APOSPC48732-5G
2'-Desoxy-2'-fluoruridin 98%
2'-Deoxy-2'-fluorouridine
Formel:
C₉H₁₁FN₂O₅ Molecular Weight: 246,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01317293 CAS-Nummer: 784-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...