2-Brom-3,4-difluoranilin 97%

Lieferant: Apollo Scientific

PC48744-25G
APOSPC48744-25GEA 119 EUR
APOSPC48744-25G
2-Brom-3,4-difluoranilin 97%
2-Brom-3,4-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,00 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035924
CAS-Nummer: 1092349-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR