(7-Fluoro-1-benzothiophen-2-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Fluoro-2-(hydroxymethyl)benzo[b]thiophene
PC48754-1G
PC48754-5G
APOSPC48754-1GEA
168
EUR
APOSPC48754-1G
APOSPC48754-5G
(7-Fluoro-1-benzothiophen-2-yl)methanol
(7-Fluoro-1-benzothiophen-2-yl)methanol
Formel:
C₉H₇FOS Molecular Weight: 182,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11841047 CAS-Nummer: 1135283-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...