1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-(ethoxycarbonylmethyl)-3-thiosemicarbazide
Lieferant: Apollo Scientific
PC4878-1G
PC4878-500MG
APOSPC4878-1GEA
977
EUR
APOSPC4878-1G
APOSPC4878-500MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-(ethoxycarbonylmethyl)-3-thiosemicarbazide
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-(ethoxycarbonylmethyl)-3-thiosemicarbazide
Formel:
C₁₁H₁₂ClF₃N₄O₂S Schmelzpunkt: 196…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975544 CAS-Nummer: 321433-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...