1-(4-Fluorophenyl)-2-[2-(methylsulphanyl)pyrimidin-4-yl]ethan-1-one 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorophenyl)-2-(2-(methylthio)pyrimidin-4-yl)ethanone

PC48790-1G PC48790-250MG
APOSPC48790-1GEA 275 EUR
APOSPC48790-1G APOSPC48790-250MG
1-(4-Fluorophenyl)-2-[2-(methylsulphanyl)pyrimidin-4-yl]ethan-1-one 95%
1-(4-Fluorophenyl)-2-[2-(methylsulphanyl)pyrimidin-4-yl]ethan-1-one
Formel: C₁₃H₁₁FN₂OS
Molecular Weight: 262,31 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18157689
CAS-Nummer: 217661-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR