Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate 97%

Lieferant: Apollo Scientific

PC4882-1G PC4882-500MG PC4882-5G
APOSPC4882-1GEA 533 EUR
APOSPC4882-1G APOSPC4882-500MG APOSPC4882-5G
Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate 97%
Ethyl-2-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetate
Formel: C₁₄H₁₇ClF₃N₃O₂
Molecular Weight: 351,76 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141695
CAS-Nummer: 338979-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR