3-Brom-2-fluor-6-iodbenzonitril 95%

Lieferant: Apollo Scientific
Achtung

PC48830-1G PC48830-5G
APOSPC48830-1GEA 161 EUR
APOSPC48830-1G APOSPC48830-5G
3-Brom-2-fluor-6-iodbenzonitril 95%
3-Brom-2-fluor-6-iodbenzonitril
Formel: C₇H₂BrFIN
Lagertemperatur: Raumtemperatur
MDL: MFCD09800805
CAS-Nummer: 217816-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR