2,5-Difluorbenzaldehyd 95%
Lieferant: Apollo Scientific
PC48832-100G
PC48832-250G
PC48832-25G
APOSPC48832-100GEA
87
EUR
APOSPC48832-100G
APOSPC48832-250G
APOSPC48832-25G
2,5-Difluorbenzaldehyd 95%
2,5-Difluorobenzaldehyde
Formel:
C₇H₄F₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010327 CAS-Nummer: 2646-90-4 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...