4-Fluorphenyloxamidsäureethylester

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-((4-fluorophenyl)amino)-2-oxoacetate, Ethyl 2-(4-fluoroanilino)-2-oxoacetate

PC4896-10G PC4896-1G PC4896-5G
APOSPC4896-10GEA 582 EUR
APOSPC4896-10G APOSPC4896-1G APOSPC4896-5G
4-Fluorphenyloxamidsäureethylester
4-Fluorphenyloxamidsäureethylester
Formel: C₁₀H₁₀FNO₃
Molecular Weight: 211.1897 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00119045
CAS-Nummer: 69065-91-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR