Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate 97%

Lieferant: Apollo Scientific

PC4901-10G PC4901-1G PC4901-5G
APOSPC4901-10GEA 800 EUR
APOSPC4901-10G APOSPC4901-1G APOSPC4901-5G
Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate 97%
Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate
Formel: C₁₅H₁₆FNO₃
Molecular Weight: 277.3 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664293
CAS-Nummer: 303139-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR