3-Bromo-2,4-difluorobenzonitrile

Lieferant: Apollo Scientific
Achtung

PC49011-1G PC49011-250MG PC49011-5G
APOSPC49011-1GEA 877 EUR
APOSPC49011-1G APOSPC49011-250MG APOSPC49011-5G
3-Bromo-2,4-difluorobenzonitrile
3-Bromo-2,4-difluorobenzonitrile
Formel: C₇H₂BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09261256
CAS-Nummer: 935534-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR