Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate 97%

Lieferant: Apollo Scientific

PC4903-1G PC4903-500MG
APOSPC4903-1GEA 799 EUR
APOSPC4903-1G APOSPC4903-500MG
Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate 97%
Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate
Formel: C₁₅H₁₃F₃N₂O₄
Molecular Weight: 342.28 g/mol
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664767
CAS-Nummer: 338761-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR