3-Methyl-7-(trifluoromethyl)pyrrolo[1,2,a]pyrazin-1-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49036-1G
PC49036-500MG
APOSPC49036-1GEA
401
EUR
APOSPC49036-1G
APOSPC49036-500MG
3-Methyl-7-(trifluoromethyl)pyrrolo[1,2,a]pyrazin-1-one 95%
3-Methyl-7-(trifluoromethyl)pyrrolo[1,2,a]pyrazin-1-one
Formel:
C₉H₇F₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD19982827 CAS-Nummer: 1025054-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...