3,3,3-Trifluoropropane-1-sulphonamide 95%

Lieferant: Apollo Scientific
Achtung

PC49037-1G PC49037-250MG
APOSPC49037-1GEA 270 EUR
APOSPC49037-1G APOSPC49037-250MG
3,3,3-Trifluoropropane-1-sulphonamide 95%
3,3,3-Trifluoropropane-1-sulphonamide
Formel: C₃H₆F₃NO₂S
Molecular Weight: 177.15 g/mol
Schmelzpunkt: 88…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08704575
CAS-Nummer: 1033906-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR