1,1,1-Trifluoro-3-methylbutan-2-ol 95%

Lieferant: Apollo Scientific
Achtung

PC49038-1G PC49038-250MG
APOSPC49038-1GEA 81.2 EUR
APOSPC49038-1G APOSPC49038-250MG
1,1,1-Trifluoro-3-methylbutan-2-ol 95%
1,1,1-Trifluoro-3-methylbutan-2-ol
Formel: C₅H₉F₃O
Molecular Weight: 142,12 g/mol
Siedepunkt: 100…102 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00462511
CAS-Nummer: 382-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR