6-Brom-8-fluorchinolin 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49040-1G
PC49040-5G
PC49040-25G
APOSPC49040-1GEA
43.5
EUR
APOSPC49040-1G
APOSPC49040-5G
APOSPC49040-25G
6-Brom-8-fluorchinolin 95%
6-Bromo-8-fluoroquinoline
Formel:
C₉H₅BrFN Molecular Weight: 226,0500 g/mol Schmelzpunkt: 103…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053791 CAS-Nummer: 220513-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...